missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)propionamide 97.0+%, TCI America™

Numéro de catalogue. H054925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H054925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H054925ML Fournisseur TCI America Code fournisseur H054925ML
Il en reste null

Identifiants chimiques

CAS 18266-55-2
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00059600
InChI Key GQKLTNAIFDFUDN-UHFFFAOYSA-N
Synonym N-Propionylethanolamine
PubChem CID 87536
IUPAC Name N-(2-hydroxyethyl)propanamide
SMILES CCC(=O)NCCO

Spécifications

Color Yellow
Quantity 25 mL
Formula Weight 117.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)propionamide