missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline Sodium Salt 98.0+%, TCI America™

Catalog No. S0827200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0827200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0827200MG Supplier TCI America Supplier No. S0827200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 82692-88-4
Color White
Molecular Formula C11H16NNaO6S
Quantity 200 mg
MDL Number MFCD00063425
Synonym 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonic Acid Sodium Salt, Sodium 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonate, HDAOS
InChI Key SVLRFMQGKVFRTB-UHFFFAOYSA-M
SMILES COC1=CC(=CC(=C1)NCC(CS(=O)(=O)[O-])O)OC.[Na+]
IUPAC Name sodium;3-(3,5-dimethoxyanilino)-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol) 313.3
PubChem CID 23666667
Formula Weight 313.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline Sodium Salt [for Biochemical Research]
Show More Show Less
TSCA No
Recommended Storage Refrigerator