missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)-N-methylaniline 98.0+%, TCI America™

Catalog No. C0872500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0872500G 500 g
C087225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0872500G Supplier TCI America Supplier No. C0872500G
Only null left

Chemical Identifiers

CAS 94-34-8
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00019856
InChI Key IXXLKTZOCSRXEM-UHFFFAOYSA-N
Synonym n-2-cyanoethyl-n-methylaniline, 3-methyl phenyl amino propanenitrile, propanenitrile, 3-methylphenylamino, n-methyl-n-2-cyanoethyl aniline, n-cyanoethyl-n-methylaniline, 3-n-methyl-n-phenylamino propionitrile, 3-n-methylanilino propionitrile, 3-n-methylanilino propanenitrile, n-.beta.-cyanoethyl-n-methylaniline
PubChem CID 66748
IUPAC Name 3-(N-methylanilino)propanenitrile
SMILES CN(CCC#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 186°C
Quantity 500 g
Formula Weight 160.22
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)-N-methylaniline
EINECSNumber (3)-0117
TSCA Yes