missing translation for 'onlineSavingsMsg'
Learn More

N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valine Methyl Ester Hydrochloride 98.0+%, TCI America™

Numéro de catalogue. C24755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C24755G 5 g
C247525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C24755G Fournisseur TCI America Code fournisseur C24755G
Il en reste null

Identifiants chimiques

CAS 482577-59-3
Molecular Formula C20H23ClN2O2
Molecular Weight (g/mol) 358.866
MDL Number MFCD08460094
InChI Key AZQXUWUZQLZNIM-FYZYNONXSA-N
PubChem CID 16744793
IUPAC Name methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride
SMILES CC(C)C(C(=O)OC)NCC1=CC=C(C=C1)C2=CC=CC=C2C#N.Cl

Spécifications

Melting Point 187°C
Color White
Quantity 5 g
Formula Weight 358.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2′-Cyanobiphenyl-4-ylmethyl)-L-valine Methyl Ester Hydrochloride