missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)acetamide 98.0+%, TCI America™

Catalog No. C14895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14895G Supplier TCI America Supplier No. C14895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7355-58-0
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.564
MDL Number MFCD00000965
InChI Key HSKNJSHFPPHTAQ-UHFFFAOYSA-N
Synonym n-2-chloroethyl acetamide, acetamide, n-2-chloroethyl, n-2-chloro-ethyl-acetamide, n-acetyl-2-chloroethylamine, n 2-chloroethyl acetamide, n-acetyl-ss-chlorathylamin, acmc-1bi05, n-2-chloroethyl-acetamide, 4-04-00-00449 beilstein handbook reference
PubChem CID 81815
IUPAC Name N-(2-chloroethyl)acetamide
SMILES CC(=O)NCCCl

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 121.56
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-(2-Chloroethyl)acetamide
RTECSNumber AB5460000
TSCA No