missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam 98.0+%, TCI America™

Catalog No. C1683500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1683500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1683500MG Supplier TCI America Supplier No. C1683500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 193202-37-8
Molecular Formula C18H19Cl2NO5S
Molecular Weight (g/mol) 432.312
MDL Number MFCD04117914
InChI Key RFABDSDXSYNRJG-UHFFFAOYSA-N
PubChem CID 44630218
SMILES CC1(C2CCC13CS(=O)(=O)N(C3C2)C(=O)C4=CC(=C(C=C4C(=O)O)Cl)Cl)C

Specifications

Color White
Quantity 500 mg
Formula Weight 432.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam