missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminophenyl)-4-[1-[2-(3-thienyl)ethyl]-1H-1,2,3-triazol-4-yl]benzamide 95.0+%, TCI America™

Catalog No. A28975MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28975MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28975MG Supplier TCI America Supplier No. A28975MG

Chemical Identifiers

CAS 1451042-18-4
Molecular Formula C21H19N5OS
Molecular Weight (g/mol) 389.48
MDL Number MFCD29089340
InChI Key LBLSLSOENGWIHL-UHFFFAOYSA-N
Synonym T 247
PubChem CID 71721946
IUPAC Name N-(2-aminophenyl)-4-{1-[2-(thiophen-3-yl)ethyl]-1H-1,2,3-triazol-4-yl}benzamide
SMILES NC1=CC=CC=C1NC(=O)C1=CC=C(C=C1)C1=CN(CCC2=CSC=C2)N=N1

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 5 mg
Formula Weight 389.48
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Aminophenyl)-4-[1-[2-(3-thienyl)ethyl]-1H-1,2,3-triazol-4-yl]benzamide
TSCA No
Recommended Storage Freezer