missing translation for 'onlineSavingsMsg'
Learn More

N-(2,3-Epoxypropyl)phthalimide, 95%

Catalog No. AC170040250
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170040250 25 g Glass bottle
AC170040100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170040250 Supplier Thermo Scientific Chemicals Supplier No. 170040250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5455-98-1 | C11H9NO3 | 203.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5455-98-1
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.20
MDL Number MFCD00005896,MFCD04973349,MFCD04973350
InChI Key DUILGEYLVHGSEE-UHFFFAOYNA-N
Synonym n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl
PubChem CID 21601
IUPAC Name 2-(oxiran-2-ylmethyl)isoindole-1,3-dione
SMILES O=C1N(CC2CO2)C(=O)C2=CC=CC=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 94.0°C to 100.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 203.2
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material N-(2, 3-Epoxypropyl)phthalimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes,on skin,or on clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for severa
EINECSNumber 226-710-2

RUO – Research Use Only