missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Thioxoethyl)glycine 97.0+%, TCI America™

Catalog No. T3312200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3312200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3312200MG Supplier TCI America Supplier No. T3312200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 412307-34-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
InChI Key MOPCBCAABYPLOC-UHFFFAOYSA-N
Synonym 2-Ethanethioamidoacetic Acid
PubChem CID 14389889
IUPAC Name 2-(ethanethioylamino)acetic acid
SMILES CC(=S)NCC(=O)O

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 133.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Thioxoethyl)glycine
TSCA No
Recommended Storage Freezer