missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)formamide 98.0+%, TCI America™

Catalog No. N10015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10015G 5 g
N100125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10015G Supplier TCI America Supplier No. N10015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6330-51-4
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD00086455
InChI Key CGRYTQQVSFZYCI-UHFFFAOYSA-N
PubChem CID 96004
IUPAC Name N-(naphthalen-1-yl)formamide
SMILES O=CNC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 140°C
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Naphthyl)formamide