missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYST2 Recombinant Rabbit Monoclonal Antibody (23GB2390)
GREENER_CHOICE

Numéro de catalogue. MA553588
Modifier l'affichage
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA553588 100 μL
MA553589 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553588 Fournisseur Invitrogen™ Code fournisseur MA553588
Il en reste null

Rabbit Recombinant Monoclonal Antibody

MYST2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene. Alternatively spliced transcript variants encoding distinct isoforms have been described.

Spécifications

Antigen MYST2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2390
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kat7
Gene Accession No. O95251, Q5SVQ0, Q810T5
Gene Alias EC 2.3.1.48; HBO1; HBOa; histone acetyltransferase binding to ORC1; Histone acetyltransferase KAT7; histone acetyltransferase MYST2; K (lysine) acetyltransferase 7; K(lysine) acetyltransferase 7; Kat7; Lysine acetyltransferase 7; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 2; MYST histone acetyltransferase 2; MYST protein 2; MYST2; MYST-2; ZC2HC7
Gene Symbols Kat7
Host Species Rabbit
Immunogen A synthesized peptide derived from human KAT7/HBO1/MYST2 (1-230AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11143, 217127, 303470
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats