missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1G Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150004
Description
MYO1G Polyclonal Antibody for Western Blot, ICC/IF, IHC
Identifiants chimiques
| MYO1G | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5SUA5 | |
| 246177 | |
| Recombinant Myosin IG (MYO1G) (Val754-Thr1018). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| MYO1G | |
| E430002D17Rik; HA2; HLA-HA2; MHAG; mHag HA-2; Minor histocompatibility antigen HA-2; Myo1g; myosin 1G; myosin IG; myosin-Ig; unconventional myosin-Ig | |
| MYO1G | |
| Primary | |
| Protein A |
Spécifications
| MYO1G | |
| Polyclonal | |
| Unconjugated | |
| MYO1G | |
| E430002D17Rik; HA2; HLA-HA2; MHAG; mHag HA-2; Minor histocompatibility antigen HA-2; Myo1g; myosin 1G; myosin IG; myosin-Ig; unconventional myosin-Ig | |
| Rabbit | |
| Protein A | |
| RUO | |
| 246177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5SUA5 | |
| MYO1G | |
| Recombinant Myosin IG (MYO1G) (Val754-Thr1018). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |