missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloma Proteins IgG1(Kappa) Isotype Control, MP Biomedicals
Suitable for use as a positive control in antibody identification screening techniques and is recommended as an ELISA coating antigen and control.
Supplier: MP Biomedicals Inc 0850327
Description
Host Species: Mouse
Isotype: IgG1 κ
Spécifications
| Myelome Protein IgG1(Kappa) | |
| MOPC 21 | |
| Mouse | |
| 1 mg |
| ELISA | |
| Purified proteins are dialyzed into 0.02M Tris, 0.15M sodium chloride, pH 8.1 | |
| 0.22μm filtered | |
| Store unopened product at -70°C. Store opened material no longer than a few days at 2° to 8°C unless sodium azide is added. For long-term storage, aliquot and freeze at -70°C. |