missing translation for 'onlineSavingsMsg'
Learn More

Mycophénolate Mofetil 98,0+%, TCI America™

Numéro de catalogue. M2387100MG
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M2387100MG 100 mg
M23871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M2387100MG Fournisseur TCI America Code fournisseur M2387100MG

Identifiants chimiques

CAS 128794-94-5
Molecular Formula C23H31NO7
Molecular Weight (g/mol) 433.501
MDL Number MFCD00867568
InChI Key RTGDFNSFWBGLEC-SYZQJQIISA-N
Synonym mycophenolate mofetil, cellcept, munoloc, myfenax, mycophenolic acid morpholinoethyl ester, tm-mmf, mycophenylate mofetil, mycophenolatemofetil, cellcept tn, unii-9242ecw6r0
PubChem CID 5281078
ChEBI CHEBI:8764
IUPAC Name 2-morpholine-4-yléthyle (E)-6-(4-hydroxy-6-méthoxy-7-méthyl-3-oxo-1H-2-benzofuran-5-yl)-4-méthylhex-4-énoate
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)OCCN3CCOCC3)OC

Spécifications

Melting Point 96°C
Color Blanc
Quantity 100 mg
Formula Weight 433.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material Mycophenolate Mofetil
RTECSNumber MP7746700
TSCA No