missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Mya 4 Reaction Station

Catalog No. 13900196
Change view
Click to view available options
For Use With (Equipment):
Pair with pumps and pH meters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13900196 Pair with pumps and pH meters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13900196 Supplier Heidolph™ Supplier No. 015620010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reaction station offering safe, precise heating and cooling with controlled operation for unattended chemistry.

  • For Use with Application:
    • Design of Experiment
    • Crystallization
    • Parallel Synthesis
    • Process Development
    • Precise Temperature Control
  • Precise temperature control
  • Magnetic or overhead stirring
  • Volume range 2 to 400 mL
  • Temperature range -30 to +180°C
  • Automation and data logging
  • Four independent reaction stations
  • Use one compact system for a range of experiments, offering a wide range of vessel sizes and styles
  • Replace inefficient, messy and unsafe oil or ice baths while saving space in the hood
  • Integrate third-party devices such as pumps, balances, and pH sensors
  • Control experiments and log results automatically
  • Software control improves safety, reduces manual errors, and allows for 24/7 unattended chemistry for improved productivity
  • A flexible discovery tool for single or parallel synthesis, lead or reaction optimization, reagent, catalyst, and solvent screening

Specifications

For Use With (Equipment) Pair with pumps and pH meters
Includes Supplied with Mya 4 Control Pad, Mya 4 to Control Pad cable, Radleys 2 GB USB stick, Mya 4 Control Pad Stylus, gas tubing: 5 m x 8.0 mm ID, reinforced coolant tubing: 5 m x 8.0 mm ID, gas flowmeter with stand: 2 to 20 L/min, coolant flowmeter with stand: 0.8 to 9 L/min, tubing clamp: 9.6 to 14 mm, blanking plugs for Mya 4 coolant connections and Mya probe and stirrer socket cover.
Product Type Reaction Station