missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide Indicator, 0.15% in Ethylene Glycol, for Calcium, Certified, LabChem™
Supplier: LabChem LC175001
| Quantity | 500 mL |
|---|---|
| Packaging | Amber Glass |
Chemical Identifiers
| C2H6O2 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| CHEBI:30742 | |
| C(CO)O |
Specifications
| Murexide Indicator | |
| 0.15% in Ethylene Glycol, for Calcium | |
| 107-21-1 , 3051-09-0 | |
| C2H6O2 | |
| Soluble in water | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 174 | |
| 284.18 | |
| Amber Glass | |
| Passes Test | |
| 1.12g/mL | |
| 500 mL | |
| Liquid |
| -13.3°C | |
| 197.8°C | |
| 99.85, 0.15 | |
| C8H8N6O6 | |
| Passes Test | |
| C(CO)O | |
| 62.068 | |
| CHEBI:30742 | |
| Certified | |
| 1.12g/mL | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
| Purple/Red | |
| 116.1°C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
EINECSNumber : 203-473-3
Recommended Storage : Room Temperature