missing translation for 'onlineSavingsMsg'
Learn More

Murexide Indicator, 0.15% in Ethylene Glycol, for Calcium, Certified, LabChem™

Catalog No. p-5389131
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC175007 125 mL Amber Glass
LC175001 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC175007 Supplier LabChem Supplier No. LC175007
Only null left

Chemical Identifiers

CAS 107-21-1 , 3051-09-0
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.3°C
Boiling Point 197.8°C
CAS Max % 99.85, 0.15
Linear Formula C8H8N6O6
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 284.18
Grade Certified
Packaging Amber Glass
Specific Gravity 1.12g/mL
Suitability Passes Test
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Density 1.12g/mL
Color Purple/Red
Quantity 125 mL
Flash Point 116.1°C
Physical Form Liquid
Chemical Name or Material Murexide Indicator, 0.15% in Ethylene Glycol, for Calcium
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 203-473-3
Recommended Storage Room Temperature