accessibility menu, dialog, popup

UserName

CAS RN 3051-09-0

IUPAC Name: ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
Synonyms: murexide
Molecular Weight (g/mol): 284.19
Molecular Formula: C8H8N6O6
InChi Key: PCCZWUCAYILEMX-UHFFFAOYSA-O
SMILES: [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1
Color 
  • (2)
  • (8)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
Grade 
  • (2)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (4)
  • (6)
  • (2)
  • (2)
  • (8)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)