Learn More
Beckman Coulter Multisizer™ 4e Smart Technology High Resolution Aperture Tubes
Smart Technology High Resolution Aperture Tubes are designed specifically for the Multisizer 4e Analyzer. For greater discrimination of particle size, high resolution tubes have an aperture thickness 120% greater than normal apertures which extends the time it takes for the particle to pass through.
$2,105.52 - $2,185.28
Spécifications
Product Type High Resolution Aperature Tube
Material Glass
For Use With (Equipment) Multisizer™ 4e Coulter Counters
voir les spécifications
Produits 4
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Diameter (Metric) Prix Quantity & Availability  
15655139
Afficher les documents Détails de la promotion
Beckman Coulter
C93978
70 μm
chaque for $2,185.28
Il en reste null
15655141
Afficher les documents Détails de la promotion
Beckman Coulter
C93979
100 μm
chaque for $2,136.96
Il en reste null
15655143
Afficher les documents Détails de la promotion
Beckman Coulter
C93981
140 μm
chaque for $2,168.42
Il en reste null
15655145
Afficher les documents Détails de la promotion
Beckman Coulter
C93983
200 μm
chaque for $2,105.52
Il en reste null
Description
Description
  • Extended measurement range (2% to 80% of aperture diameter) leads to less frequent aperture changes for more analysis options
  • A wide range of apertures is available with diameters ranging from 10 to 2,000 μm
  • Specifically designed for the Multisizer 4e or Multisizer 4 Coulter Counters
  • 100 μm ST Aperture Tube is standard with the Multisizer 4e Coulter Counter
  • Compatible with ST Beakers and Accuvette STs
  • Convenient tubes ensure accurate, and consistent results
  • For greater discrimination of particle size, high resolution tubes have an aperture thickness 120% greater than normal apertures which extends the time the particle takes to pass through
  • Size and serial number information is included on a bar code on the tube for automatic data entry via instrument bar code reader
Spécifications
Spécifications
High Resolution Aperature Tube
Glass
Multisizer™ 4e Coulter Counters
Videos
FDS