Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
Automated reagent dispenser for 6- to 1536-well microplates.
$8000.42 - $13993.97
Specifications
| For Use With (Equipment) | MultiFlo FX Microplate Dispenser |
|---|---|
| Quantity | 1/Cs. |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|
11-120-643
|
Agilent BioTek
7210010 |
Peristaltic Pump Dispenser Module |
Each for $8,000.42
|
|
|||||
|
11-120-644
|
Agilent BioTek
7210008 |
Dual Syringe Drive Dispenser (Auto) |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
11-120-645
|
Agilent BioTek
7210009 |
Dual Syringe Drive Dispenser (Non-Auto) |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Automated reagent dispenser for 6- to 1536-well microplates
- Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
- RAD technology for dispensing to individual wells
- Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
- For benchtop and integrated automation workflows
- Bulk reagent dispensing
- Cell seeding
- Cell washing /media exchange
- Bead washing
Specifications
| MultiFlo FX Microplate Dispenser | |
| 1/Cs. |