Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
Automated reagent dispenser for 6- to 1536-well microplates.
Supplier:  Agilent BioTek 7210009
 View more versions of this product
Catalog No. 11-120-645
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Automated reagent dispenser for 6- to 1536-well microplates
  • Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
  • RAD technology for dispensing to individual wells
  • Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
  • For benchtop and integrated automation workflows
  • Bulk reagent dispensing
  • Cell seeding
  • Cell washing /media exchange
  • Bead washing
Specifications
Specifications
Dual Syringe Drive Dispenser (Non-Auto)
MultiFlo FX Microplate Dispenser
1/Cs.
Product Suggestions
Product Suggestions