Learn More
Agilent BioTek MultiFlo FX Microplate Dispenser Modules
Automated reagent dispenser for 6- to 1536-well microplates.
Supplier: Agilent BioTek 7210010
Description
- Automated reagent dispenser for 6- to 1536-well microplates
- Parallel Dispense technology allows up to four independent reagents to be dispensed in parallel without potential carryover
- RAD technology for dispensing to individual wells
- Cell friendly angled dispense and/or wash tubes won't disrupt cell layers
Applications:
- For benchtop and integrated automation workflows
- Bulk reagent dispensing
- Cell seeding
- Cell washing /media exchange
- Bead washing
Specifications
| Peristaltic Pump Dispenser Module | |
| MultiFlo FX Microplate Dispenser | |
| Plastic tip cassette | |
| 1/Cs. |