missing translation for 'onlineSavingsMsg'
Learn More

Multi Ion anion IC standard solution, Specpure™ (Waters) in H{2}O

Catalog No. AA35564AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35564AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35564AE Supplier Thermo Scientific Chemicals Supplier No. 035564AE

 

Multi Ion anion IC standard solution is a high purity starting material, which is a certified primary reference material for Ion Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Multi Ion anion IC standard solution
Name Note Specpure™ (Waters) in H2O
Physical Form Liquid
MDL Number MFCD00144814
Solubility Miscible with water
Quantity 100 mL
Grade Specpure

RUO – Research Use Only