Learn More
Thermo Scientific™ Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Catalog No. 14385374
Change view
Click to view available options
Type:
4-position Carousel Base
8-position Carousel
Rect Cell 4-position Carousel
Holds:
4 x 10mm Pathlength Cells
4 x 50mm PL Rectangular Cells
8 x 10mm Pathlength Cells
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Holds
14385374 8-position Carousel 8 x 10mm Pathlength Cells
14380455 Rect Cell 4-position Carousel 4 x 50mm PL Rectangular Cells
14385375 4-position Carousel Base 4 x 10mm Pathlength Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14385374 Supplier Thermo Scientific™ Supplier No. 840303400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Speed up your analysis with high-throughput measurements.
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
Specifications
Compatibility Compatible with 10mm pathlength cells only. Compatible with 4.5mL cell and semi-micro cells with windows ≥ 3mm width (≥ 1000μL). Please note: Not compatible with microcells (less than 1000μL in volume or with GENESYS 30/40/50 spectrophotometers).
Type 8-position Carousel
For Use With (Equipment) GENESYS G5
Holds 8 x 10mm Pathlength Cells