missing translation for 'onlineSavingsMsg'
Learn More

Mucobromic Acid 99.0+%, TCI America™

Catalog No. M0471500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0471500G 500 g
M047125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0471500G Supplier TCI America Supplier No. M0471500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 488-11-9
Molecular Formula C4HBr2O3-
Molecular Weight (g/mol) 256.857
MDL Number MFCD00063745
InChI Key NCNYEGJDGNOYJX-IHWYPQMZSA-M
Synonym mucobromic acid, z-2,3-dibromo-4-oxo-2-butenoate, z-2,3-dibromo-4-oxobut-2-enoate, 2z-2,3-dibromo-4-oxobut-2-enoate, z-2,3-bis bromanyl-4-oxidanylidene-but-2-enoate
PubChem CID 6994897
IUPAC Name (Z)-2,3-dibromo-4-oxobut-2-enoate
SMILES C(=O)C(=C(C(=O)[O-])Br)Br

Specifications

Melting Point 124°C
Color White
Quantity 500 g
UN Number 3261
Formula Weight 257.87
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mucobromic Acid
EINECSNumber (2)-1165
TSCA Yes