missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ MTS™ Doxycycline [DXT] 0.016-256 μg/mL
MIC Test Strips are a quantitative assay for determining the Minimum Inhibitory Concentration (MIC) of antimicrobial agents against microorganisms to indicate appropriate patient treatment and for identifying resistance patterns.
$582.89
Spécifications
| Antimicrobial Agents | Doxycycline |
|---|---|
| Certifications/Compliance | FDA Cleared |
| Concentration | 0.016 to 256 μg/mL |
| Content And Storage | -20°C or 2°C to 8°C until the given expiry date |
| Description | Liofilchem™ MTS™ Doxycycline |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging Type | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging Type | Prix | Quantity & Availability | ||||
22777990
|
Liofilchem™
92156 |
30/Pk. | Individually Packed |
paquet de 30 chaque for $582.89
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Spécifications
| Doxycycline | |
| 0.016 to 256 μg/mL | |
| Liofilchem™ MTS™ Doxycycline | |
| MIC Test Strip |
| FDA Cleared | |
| -20°C or 2°C to 8°C until the given expiry date | |
| DXT | |
| Antimicrobial Susceptibility Testing, For determining the MIC of antimicrobial agents against microorganisms |
Non-hazardous according to current legislation, but falls within the specific field of application where a safety data sheet must be supplied because it can cause phenomena of sensitization in sensitive subjects if it comes into contact with the skin.