missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTMR1 Polyclonal Antibody

Numéro de catalogue. PIPA542117
Modifier l'affichage
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA542117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA542117 Fournisseur Invitrogen™ Code fournisseur PA542117

Rabbit Polyclonal Antibody

Peptide sequence: KEDVYTKTIS LWSYINSQLD EFSNPFFVNY ENHVLYPVAS LSHLELWVNY Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

MTMR1 is a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases.This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined.

Spécifications

Antigène MTMR1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 2% sucrose and 0.09% sodium azide
Gène MTMR1
Numéro d’acquisition du gène Q13613
Alias du gène AW049210; LOW QUALITY PROTEIN: myotubularin-related protein 1; Mtmr1; myotubularin related protein 1; myotubularin-related protein 1; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase
Symboles de gène MTMR1
Espèces hôtes Rabbit
Immunogène Synthetic peptide directed towards the C-terminal of human MTMR1 (aa 551-600).
Méthode de purification Protein A
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 8776
Espèces cibles Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats