Learn More
MilliporeSigma mPAGE™ Mini Gel Tank
Tank for electrophoresis of precast gels or hand cast polyacrylamide gels
$887.42 - $1130.00
Specifications
For Use With (Application) Gel Electrophoresis
For Use With (Equipment) Gels cast with mPage™, Gel Caster Other precast gels with cassette dimension 8 x 10 cm mPAGE™, Bis-Tris Precast Gels mPAGE™, Mini Wet Transfer System
Type Gel Tank
Max. Voltage 300 V
View More Specs
Products 2
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
11-580-593
View Documents
MilliporeSigma
MGT2
2 Mini-gels
Each for $887.42
Only null left
11-580-594
View Documents
MilliporeSigma
MGT4
4 Gels
Each for $1,130.00
Only null left
Description
Description
  • The mPAGE™ Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels.
  • The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE™ Gel Caster and mPAGE™ Precast Gels.
  • The mPAGE™ Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.
Specifications
Specifications
Gel Electrophoresis
Gels cast with mPage™, Gel Caster Other precast gels with cassette dimension 8 x 10 cm mPAGE™, Bis-Tris Precast Gels mPAGE™, Mini Wet Transfer System
Gel Tank
300 V
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'