Learn More
MilliporeSigma mPAGE™ Mini Gel Tank
Description
- The mPAGE™ Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels.
- The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE™ Gel Caster and mPAGE™ Precast Gels.
- The mPAGE™ Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.
Specifications
Specifications
| Capacity | 4 Gels |
| For Use With (Application) | Gel Electrophoresis |
| For Use With (Equipment) | Gels cast with mPage™, Gel Caster Other precast gels with cassette dimension 8 x 10 cm mPAGE™, Bis-Tris Precast Gels mPAGE™, Mini Wet Transfer System |
| Type | Gel Tank |
| Max. Voltage | 300 V |