Learn More
MilliporeSigma mPAGE™ Mini Gel Tank
Catalog No. 11580593
Change view
Click to view available options
Capacity:
2 Mini-gels
4 Gels
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
11580593 2 Mini-gels
11580594 4 Gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11580593 Supplier MilliporeSigma Supplier No. MGT2
Only null left
Tank for electrophoresis of precast gels or hand cast polyacrylamide gels
  • The mPAGE™ Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels.
  • The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE™ Gel Caster and mPAGE™ Precast Gels.
  • The mPAGE™ Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.
Specifications
Capacity 2 Mini-gels
For Use With (Application) Gel Electrophoresis
For Use With (Equipment) Gels cast with mPage™, Gel Caster Other precast gels with cassette dimension 8 x 10 cm mPAGE™, Bis-Tris Precast Gels mPAGE™, Mini Wet Transfer System
Type Gel Tank
Max. Voltage 300 V