missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MP Magnetic Rack 96

Catalog No. MP116570429
Change view
Click to view available options
Color:
Gold
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
MP116570429 Gold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116570429 Supplier MP Biomedicals Supplier No. 116570429
Only null left

MP Magnetic Rack 96 is a novel tool designed for fast and easy manual extraction of nucleic acids from standard 96-well plates using a magnetic bead extraction method.

  • Convenient and easy-to-handle for high-throughput manual nucleic acid extractions
  • 9 rows of 13 magnet pins ensures fast and efficient magnetic bead separation from solutions
  • It is commonly used in combination with MagBeads FastDNA and FastRNA kits from MP Biomedicals
  • The magnetic stand contains a total of 117 strong pin magnets for fast and efficient magnetic bead separation from solutions when using 96-deep well plates with either U- or V-bottom wells.
  • The magnetic force generated by the stand pulls magnetic beads from the solution in just a few minutes, thus decreasing the chance of sample loss.

Specifications

Autoclavable Not Autoclavable
Color Gold
Holds Microplates (Standard Size Flat)
Positions 96 positions
Shape Rectangular
Stackable No
Sterility Non-sterile
Product Type Magnetic Rack
Shelves None
Array 9 x 13