missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Goat, Purified, 2mL

Catalog No. ICN55984
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55984 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55984 Supplier MP Biomedicals™ Supplier No. 0855984

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Whole Goat Serum with buffer salts

Specifications

Content And Storage 2° to 8°C
Species Goat
Product Type Goat Serum
Quantity 2 mL
Sterilization Method 0.2 μm filtration
Formulation Delipidated, lyophilized from PBS with Azide
For Use With (Application) Blocking assays, immunohistochemical, immunocytochemical, immunofluorescent applications
Lyophilized Yes
Protein Content 63.3 mg/mL