missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ribonuclease A, From Bovine Pancreas
Aggregates can be converted to monomers.
$199.08 - $1103.76
Specifications
| Concentration | ≥50 kU/mg |
|---|---|
| Color | White |
| Purity | >70% |
| pH | 7 to 7.5 |
| For Use With (Application) | Used to remove RNA from DNA plasmid preparations and Protein samples |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10107680
|
MP Biomedicals™
0210107680 |
100 mg |
Each for $199.08
|
|
|||||
|
ICN10107683
|
MP Biomedicals™
0210107683 |
250 mg |
Each for $402.11
|
|
|||||
|
ICN10107691
|
MP Biomedicals™
0210107691 |
1 g |
Each for $1,103.76
|
|
|||||
Description
Ribonuclease A, [9001-99-4]
- RNase A
- From Bovine Pancreas
- Prepared from aggregate-free RNase
- Lyophilized
- Activity: 50 Kunitz units/mg.
Specifications
| ≥50 kU/mg | |
| >70% | |
| Used to remove RNA from DNA plasmid preparations and Protein samples | |
| Bovine pancreas | |
| 13700 | |
| Lyophilized Powder |
| White | |
| 7 to 7.5 | |
| 0°C | |
| Ribonuclease A | |
| Heavy Metal ions and is competitively inhibited by DNA |