missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ribonuclease A, From Bovine Pancreas
Aggregates can be converted to monomers.
Supplier: MP Biomedicals™ 0210107683
Description
Ribonuclease A, [9001-99-4]
- RNase A
- From Bovine Pancreas
- Prepared from aggregate-free RNase
- Lyophilized
- Activity: 50 Kunitz units/mg.
Specifications
| ≥50 kU/mg | |
| >70% | |
| Used to remove RNA from DNA plasmid preparations and Protein samples | |
| Bovine pancreas | |
| Ribonuclease A | |
| Heavy Metal ions and is competitively inhibited by DNA |
| White | |
| 7 to 7.5 | |
| 0°C | |
| 250 mg | |
| 13700 | |
| Lyophilized Powder |