missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Serum, Rabbit

Catalog No. ICN2941149
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2941149 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2941149 Supplier MP Biomedicals™ Supplier No. 092941149

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supports cultures of Borrelia burgdorferi, the causative agent of Lyme disease.

This serum is derived from the blood of healthy adult animals. Rabbit serum is used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease.

Specifications

Certifications/Compliance U.S. regulations
Color Dark Brown
Content And Storage -20°C
Species Rabbit
Form Frozen
Product Type Rabbit Serum
Quantity 100 mL
Concentration 4 to 6.6 g/dL
Sterility Sterile
Sterilization Method Filtration
For Use With (Application) Used to support cultures of Borrelia burgdorferi, the causative agent of Lyme disease
Hemoglobin Concentration ≤33 mg/dL
Mycoplasma Detected No
Osmolality 268 to 326 mOsm/kg
pH 7.4 to 7.9
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.