missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein Precipitate Solution (PPS)

Catalog No. 116540403
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116540403 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 116540403 Supplier MP Biomedicals™ Supplier No. 116540403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Component of FastDNA™ SPIN Kit for Soil

PPS is a lysis solution component included in the FastDNA and the FastDNA Spin Kits. Used as a stabilization agent during sample lysis of primarily plant tissue, but can also be used for other sample types that are high in inhibitory compounds. PPS is a protein precipitation solution used in conjunction with a the CLS-VF lysing buffer for homogenization of plant tissue for the subsequent extraction and purification of nucliec acids.

Specifications

Color Clear
Content And Storage Stable at room temperature. Shelf life may be extended by storing at 4°C
For Use With (Equipment) FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-1
For Use With (Application) Nucleic Acid Isolation & Purification
pH 6.18
Product Type PPS
Quantity 25 mL
Form Liquid