missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sodium Iodide (NaI) Solution

Catalog No. MP111001601
Change view
Click to view available options
Quantity:
1 x 60 mL
1 x 125 mL
2 x 125 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111001601 2 x 125 mL
MP111001201 1 x 60 mL
MP111001401 1 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP111001601 Supplier MP Biomedicals™ Supplier No. 111001601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Iodide (NaI) Solution dissolves agarose gels and helps in binding the DNA to the Glassmilk™ binding matrix.

  • Less than 15% DNA 1 kb and above is left unbound in NaI.
  • NaI (Sodium Iodide) is supplied with GeneClean™ kits and helps in binding the DNA to the GLASSMILK binding matrix.
  • Melts 0.1 g gel slice in 5 minutes at 55°C.

Specifications

CAS 7681-82-5
Concentration 6 M
Content And Storage 15°C to 30°C
Format Bottle
Isolation Technology Glass Milk
Formula Weight 149.89
For Use With (Application) Nucleic Acid Isolation and Purification
Molecular Formula NaI
Melting Point 661°C
pH 7.0 to 7.4
Product Type NaI
Quantity 2 x 125 mL
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.