Learn More
MP Biomedicals™ Lysing Matrix Y, 96-Tube Rack
Lysing Matrix Y, 96-Tube Rack is pre-filled with 0.5 mm diameter yttria-stabilized zirconium oxide beads and used to lyse yeast, fungi and algae.
Supplier:  MP Biomedicals™ 116960010
 View more versions of this product
Catalog No. MP116960010
Only null left
Description
Description
  • Each 96 deep well rack comes with 1.2mL cluster-strip tube and cap assemblies.
  • Each tube is pre-filled with 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads.
  • Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimizes shearing.
  • These Lysing Matrix racks are optimized for the FastPrep™-96 Instrument and are included in the FastDNA™-96 Soil Microbe DNA Kit, the FastDNA™-96 Fungal/Bacterial DNA Kit, and the FastDNA™-96 Fecal DNA Kit
Specifications
Specifications
Yttria-stabilized Zirconium Oxide
Yeast, Fungi, Algae
Nucleic Acid Isolation, Homogenization
96
1.2 mL
FastPrep-96
1.2 mL Cluster-strip Tube and Cap Assemblies Pre-filled with Beads
10 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.