Learn More
MP Biomedicals™ Lysing Matrix Y, 96-Tube Rack
Lysing Matrix Y, 96-Tube Rack is pre-filled with 0.5 mm diameter yttria-stabilized zirconium oxide beads and used to lyse yeast, fungi and algae.
Supplier:  MP Biomedicals™ 116960001
 View more versions of this product
Catalog No. MP116960001
Only null left
Description
Description
  • Each 96 deep well rack comes with 1.2mL cluster-strip tube and cap assemblies.
  • Each tube is pre-filled with 0.5 mm diameter Yttria-Stabilized Zirconium Oxide beads.
  • Y2O3 + ZrO2 has high density and has extremely high hardness making this matrix ideal for cracking of very tough cell walls, while it's smooth surface minimizes shearing.
  • These Lysing Matrix racks are optimized for the FastPrep™-96 Instrument and are included in the FastDNA™-96 Soil Microbe DNA Kit, the FastDNA™-96 Fungal/Bacterial DNA Kit, and the FastDNA™-96 Fecal DNA Kit
Specifications
Specifications
Yttria-stabilized Zirconium Oxide
Yeast, Fungi, Algae
Nucleic Acid Isolation, Homogenization
96
1.2 mL
FastPrep-96
1.2 mL Cluster-strip Tube and Cap Assemblies Pre-filled with Beads
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.