missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix F, 2 mL Tubes
Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
$259.98 - $2417.10
Specifications
| Sample Type | Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold |
|---|---|
| DNase | Dnase-free |
| For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
| Matrix | Lysing matrix F |
| Molecular Formula | Aluminum Oxide, Al2O3; Silicon Carbide, SiC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP116915050
|
MP Biomedicals™
116915050 |
50 x 2 mL Tubes |
Each for $259.98
|
|
|||||
|
MP116915100
|
MP Biomedicals™
116915100 |
100 x 2 mL Tubes |
Each for $509.52
|
|
|||||
|
MP116915500
|
MP Biomedicals™
116915500 |
500 x 2 mL Tubes |
Each for $2,417.10
|
|
|||||
Description
- Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
- With white caps
- Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
- Nucleic Acid Isolation and Purification
Specifications
| Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold | |
| Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
| Aluminum Oxide, Al2O3; Silicon Carbide, SiC |
| Dnase-free | |
| Lysing matrix F | |
| RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.