missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix F, 2 mL Tubes
Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
Supplier: MP Biomedicals™ 116915500
Description
- Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
- With white caps
- Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
- Nucleic Acid Isolation and Purification
Specifications
| 2 mL | |
| Dnase-free | |
| Lysing matrix F | |
| 500 x 2 mL Tubes |
| Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold | |
| Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis | |
| Aluminum Oxide, Al2O3; Silicon Carbide, SiC | |
| RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.