missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hyaluronidase, Bovine Testes
Bovine Testes
$145.92 - $1135.60
Specifications
| Concentration | ≥300 U/mg |
|---|---|
| Color | Tan |
| pH | 4.5 to 6 |
| For Use With (Application) | Drug dispersion and delivery |
| Content And Storage | -20°C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10074080
|
MP Biomedicals™
0210074080 |
100 mg |
Each for $145.92
|
|
|||||
|
ICN10074091
|
MP Biomedicals™
0210074091 |
1 g |
Each for $1,135.60
|
|
|||||
|
ICN10074090
|
MP Biomedicals™
0210074090 |
500 mg |
Each for $296.33
|
|
|||||
Description
Hyaluronidase, [37326-33-3]
- Hyaluronate Glycanohydrolase
- Activity: 300 USP units/mg min.
- Lyophilized powder
Specifications
| ≥300 U/mg | |
| 4.5 to 6 | |
| -20°C | |
| Hyaluronidase | |
| Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+ |
| Tan | |
| Drug dispersion and delivery | |
| Bovine testes | |
| ≈55 | |
| Lyophilized Powder |