missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hyaluronidase, Bovine Testes
Bovine Testes
Supplier: MP Biomedicals™ 0210074090
Description
Hyaluronidase, [37326-33-3]
- Hyaluronate Glycanohydrolase
- Activity: 300 USP units/mg min.
- Lyophilized powder
Specifications
| ≥300 U/mg | |
| 4.5 to 6 | |
| -20°C | |
| 500 mg | |
| ≈55 | |
| Lyophilized Powder |
| Tan | |
| Drug dispersion and delivery | |
| Bovine testes | |
| Hyaluronidase | |
| Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+ |