missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Serum, Goat, Heat Inactivated

Catalog No. ICN2939249
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2939249 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2939249 Supplier MP Biomedicals™ Supplier No. 092939249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Goat Serum is used as a supplement to cell culture media.

Goat Serum is used as a supplement to cell culture media. Goat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health.

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Goat
Product Type Goat Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
Heat Inactivated Yes

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.