missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastRNA™ Pro Soil-Indirect Kit

Catalog No. MP116075050
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116075050 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116075050 Supplier MP Biomedicals™ Supplier No. 116075050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FastRNA™ Pro Soil-Indirect Kit is used to isolate total RNA from organic material found in soil samples. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.

  • This kit allows for the sample to be slurried with water or buffer in order to further separate biomass from the soil or environmental matrix, and the resulting supernatant is then filtered, lysed, and purified through the rest of the protocol.
  • Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.

Specifications

Sample Type Feces, Soil, Wastewater, Sludge
No. of Reactions 50 Preps
Product Type Kit
Quantity 2 mL
Sample Size Up to 500 mg per prep
Sample Source Microorganisms

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.