Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
Supplier:  MP Biomedicals™ 0219102305
 View more versions of this product
Catalog No. ICN19102305
Only null left
Explore available promotions
Description
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
Specifications
Specifications
15° to 30°C
Amicase
5
5 lb.
Product Suggestions
Product Suggestions