missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Carbonic Anhydrase

Supplier:  MP Biomedicals™ 0215387925

 View more versions of this product

Catalog No. ICN15387925

Only null left
Explore available promotions

Description

Description

  • Activity 2000units/mg

Specifications

Specifications

>2000 U/mg
Molecular biology, cell biology
Bovine erythrocytes
Carbonic Anhydrase
Lyophilized Powder
Tan
2°C to 8°C
25 mg
30000
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions