missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Arginase from Bovine Liver
Major degradative enzyme for arginine that converts arginine to ornithine
Supplier: MP Biomedicals™ 0210027710
Description
The enzyme participates in the Krebs-Henseleit urea cycle. It is most highly concentrated in mammalian liver and also present in mammary glands.
- Synonyms: L-arginine amidinase, L-arginine amidino-hydrolase
- CAS Number: 9000-96-8
- EC Number: 232-570-3
- MDL Number: MFCD00081368
- Format: Brown powder
- Specific Activity: ∽20U/mg
- Extinction Coefficient E1%: 278nm
- pH: Optimum pH: 10 (Mn2+ activated)
- Dissolves readily at 1mg/mL in 0.05M Manganese maleate (pH 7.0) to give clear pale brown solution
- Soluble in distilled water or diluted buffer
Specifications
| 1 mg/mL | |
| 10 | |
| -20°C | |
| 10 kU | |
| 115000 to 120000 | |
| Powder |
| Brown | |
| Converts Arginine to Ornithine, deficiency is associated with spasticity and motor dysfunction | |
| Bovine liver | |
| Arginase | |
| Hg2+, Ag2+, Zn2+ |