missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Arginase from Bovine Liver

Catalog No. p-4605316
Change view
Click to view available options
Quantity:
10 kU
3 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10027710 10 kU
ICN10027703 3 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10027710 Supplier MP Biomedicals™ Supplier No. 0210027710

Major degradative enzyme for arginine that converts arginine to ornithine

The enzyme participates in the Krebs-Henseleit urea cycle. It is most highly concentrated in mammalian liver and also present in mammary glands.

  • Synonyms: L-arginine amidinase, L-arginine amidino-hydrolase
  • CAS Number: 9000-96-8
  • EC Number: 232-570-3
  • MDL Number: MFCD00081368
  • Format: Brown powder
  • Specific Activity: ∽20U/mg
  • Extinction Coefficient E1%: 278nm
  • pH: Optimum pH: 10 (Mn2+ activated)
  • Dissolves readily at 1mg/mL in 0.05M Manganese maleate (pH 7.0) to give clear pale brown solution
  • Soluble in distilled water or diluted buffer

Specifications

Concentration 1 mg/mL
Color Brown
pH 10
For Use With (Application) Converts Arginine to Ornithine, deficiency is associated with spasticity and motor dysfunction
Content And Storage -20°C
Source Bovine liver
Quantity 10 kU
Product Type Arginase
Molecular Weight (g/mol) 115000 to 120000
Inhibitors Hg2+, Ag2+, Zn2+
Form Powder
Show More Show Less