missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Acid Phosphatase, Wheat Germ
Manufacturer: MP Biomedicals™ 0215387690
Description
- Activity 20units/mg
Specifications
Lyophilized Powder | |
Brown | |
4 to 7 | |
Wheat germ | |
Acid Phosphatase |
20 U/mg | |
90% | |
Catalyzes the hydrolysis of Monoesters and Anhydrides of Phosphoric Acid | |
500 mg | |
58 |